Learn More
Labcon ZAP™ 10 μL Extra Long Aerosol Filter Pipette Tips
Refillable autoclavable racks
Marca: Labcon 1191-965-006-9
Descrizione
- Pipette tip
- Aerosol Filter
- Patented robotically inserted endotoxin free filter
- Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Made in USA
Specifica
10 μL | |
Sterile | |
Purple | |
1.87 in. | |
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Liquid Handling | |
UltraFine | |
96 tips / rack | |
Yes | |
Universal Fit | |
ZAP |
Con filtro | |
Natural | |
Not Autoclavable | |
47.6 mm | |
Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) | |
In rack | |
96 tips/rack; 10 racks/Pk.; 10 Pk./case | |
RT | |
Single-use | |
No | |
Polypropylene Resin |