missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551178
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total ZCCHC9 protein.
ZCCHC9 Polyclonal antibody specifically detects ZCCHC9 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
ZCCHC9 | |
Polyclonal | |
Unconjugated | |
ZCCHC9 | |
PPP1R41; protein phosphatase 1, regulatory subunit 41; ZCCHC9; Zinc finger CCHC domain-containing protein 9; zinc finger CCHC-type containing 9; zinc finger, CCHC domain containing 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84240 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N567 | |
ZCCHC9 | |
Fusion protein of human ZCCHC9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |