missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenon™ pHrodo™ iFL IgG Labeling Reagents

Codice prodotto. 30535017
Cambia vista
Click to view available options
Quantità:
1.2 mL
Specie:
Human
Mouse
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo Red
Colore:
Deep Red
Green
Red
Dimensione della confezione:
1.20mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Specie Label or Dye Colore Dimensione della confezione
30535017 1.2 mL Mouse pHrodo Red Red 1.20mL
30534651 1.2 mL Human pHrodo Deep Red Deep Red 1.20mL
30535648 1.2 mL Mouse pHrodo Deep Red Deep Red 1.20mL
30535977 1.2 mL Mouse pHrodo Green Green 1.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30535017 Fornitore Invitrogen™ N. del fornitore Z25626
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.

Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Store in refrigerator at 2°C to 8°C.

TRUSTED_SUSTAINABILITY

Specifica

Da utilizzare con (applicazione) To screen for antibody internalization
Product Type Labeling Reagent
Quantità 1.2 mL
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Metodo di rilevamento Fluorescence
Excitation/Emission 560/585 nm
Da utilizzare con (apparecchiatura) Flow Cytometer, High Content Analysis, IncuCyte, Fluorescence Microscope
Labeling Method Rapid, no wash
Linea di prodotti Labeling Kits
Solubility Aqueous Buffer
Label or Dye pHrodo Red
Colore Red
Labeling Target Antibodies
Specie Mouse
Tipo di etichetta Mouse IgG Fc domain
Vedi altri risultati Mostra meno risultati

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.