missing translation for 'onlineSavingsMsg'
Learn More

ZFP36L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12306-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899943

  • 140.00€ / 20µL
Spedizione stimata: 03-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The gene is well conserved across species and has a promoter that contains motifs seen in other early-response genes. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors.
Specifica

Specifica

ZFP36L1
Polyclonal
Unconjugated
ZFP36L1
Berg36, BRF1, Butyrate response factor 1, cMG1, EGF response factor 1, ERF 1, ERF1, Protein TIS11B, RNF162B, TIS11B, ZFP36L1
Rabbit
Antigen Affinity Chromatography
RUO
12192, 29344, 677
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17431, P23950, Q07352
ZFP36L1
ZFP36L1 Fusion Protein Ag2952
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali