missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7082677
34.20 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 m
25 m
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Zinc wire
Nota nome 1.0mm (0.04 in.) dia
Densità ≈5.60g/m
Note percentuale saggio (metals basis)
Quantità 1 m
Percent Purity 99.95%
Odore Odorless

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'