Learn More
ZNF106 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562943
Dettagli aggiuntivi : Peso : 0.02000kg
Description
Immunogen sequence: RSGWHKGVAG GSSTWFHNHS NSGGGWLSNS GAVDWNHNGT GRNSSWLSEG TGGFSSWHMN NSNGNWKSSV RSTNNWNYSG PGDKFQPGRN RNSNCQMEDM TMLWNKKSNK SNKYSHDRYN WQRQENDKLG TVATYRGPSE GFTSD Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
ZNF106 Polyclonal antibody specifically detects ZNF106 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spécification
ZNF106 | |
Polyclonal | |
Unconjugated | |
ZNF106 | |
Cd-1; D2Dcr28; H3a; H3a minor histocompatibility antigen; Sh3bp3; SH3-domain binding protein 3; sirm; ZFP106; zfp-106; zinc finger protein 106; Zinc finger protein 474; Znf106; ZNF474 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64397 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2Y7 | |
ZNF106 | |
Recombinant protein corresponding to Human ZNF106 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only