missing translation for 'onlineSavingsMsg'
Learn More

ZNF2 Recombinant Protein, Abnova

Marca:  Abnova H00007549-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16161942

  • 302.00€ / 10µg


Spedizione stimata
24-11-2020
Aggiungi al carrello

Descrizione

Descrizione

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH05068
Solution
7549
ZNF2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZNF2
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.86
8
Glutathione Sepharose 4 Fast Flow
10 ug
MGTEKQSPSGETRKKSLSRDKGLRRRSALSREILTKERHQECSDCGKTFFDHSSLTRHQRTHTGEKPYDCRECGKAFSHRSSLSRHLMSHTGESPYECSVCSKAFFDRSSLTVHQRIHTGEKPFQCNECGKAFFDRSSLTRHQRIHTGESPYECHQCGKAFSQKSILTRHQLIHTGRKPYECNECGKAFYGVSSLNRHQKAHAGDPRYQCNECGKAFFDRSSLTQHQKIHTGDKPHECSECGKAFSQRCRLTRHQRVHTGEKPFECTVCGKVFSSKSSVIQHQRRYAKQGID
A1-5|ZNF661|Zfp661
ZNF2
Wheat Germ (in vitro)
GST
Videos
Documenti