missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF221 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563722
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MISPSLELLH SGLCKFPEVE GKMTTFKEA Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 34%.
ZNF221 may be involved in transcriptional regulation.Specifica
ZNF221 | |
Polyclonal | |
Unconjugated | |
ZNF221 | |
Zinc finger protein 221; ZNF221 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7638 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK13 | |
ZNF221 | |
Recombinant protein corresponding to Human ZNF221 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only