missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF80 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA564225
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LEHRKDFGYH SAFAQQSKIH SGGKNL Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 42%.
ZNF80 may be involved in transcriptional regulation.Specifica
ZNF80 | |
Polyclonal | |
Unconjugated | |
ZNF80 | |
pT17; zinc finger protein 80; ZNF80; ZNFpT17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51504 | |
ZNF80 | |
Recombinant protein corresponding to Human ZNF80 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only