Learn More
Zofenoprilat Sodium Salt (90%), Zofenoprilat Sodium Salt (90%), TRC
Product Code. 17950495
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950495 1 mg 1mg
30136178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17950495 Supplier Toronto Research Chemicals Supplier No. TRCZ5870501MG
 Restricted Item
Spedizione stimata: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C15H18NNaO3S2
Molecular Weight (g/mol) 347.43
Synonym (4S)-1-[(2S)-3-Mercapto-2-methyl-1-oxopropyl]-4-(phenylthio)-L-proline Sodium, [1(R*),2α,4α]-1-(3-Mercapto-2-methyl-1-oxopropyl)-4-(phenylthio)-L-proline Sodium;
IUPAC Name sodium;(2S,4S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]-4-phenylsulfanylpyrrolidine-2-carboxylate
SMILES O=C([C@H](C)CS)N1[C@@H](C(O[Na])=O)C[C@H](SC2=CC=CC=C2)C1
Specifications
CAS 1329569-13-2
Quantity 1 mg
InChI Formula InChI=1S/C15H19NO3S2.Na/c1-10(9-20)14(17)16-8-12(7-13(16)15(18)19)21-11-5-3-2-4-6-11;/h2-6,10,12-13,20H,7-9H2,1H3,(H,18,19);/q;+1/p-1/t10-,12+,13+;/m1./s1
Formula Weight 347.0626
Chemical Name or Material Zofenoprilat Sodium Salt