missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zolpidem tartrate 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Materiale o nome chimico | Zolpidem Tartrate |
| CAS | 99294-93-6 |
| Nomi componenti o analita | Zolpidem Tartrate |
| Shipping Condition | +20°C |
| Formula molecolare | 2 C19 H21 N3 O . C4 H6 O6 |
| Concentrazione | 100 μg/mL |
| Certificazione/Conformità | ISO 17034 |
| Sinonimo | Zolpidem Tartrate |
| InChI Formula | InChI=1S/2C19H21N3O.C4H6O6/c2*1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19;5-1(3(7)8)2(6)4(9)10/h2*5-10,12H,11H2,1-4H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1 |
| SMILES | CN(C)C(=O)Cc1c(nc2ccc(C)cn12)c3ccc(C)cc3.CN(C)C(=O)Cc4c(nc5ccc(C)cn45)c6ccc(C)cc6.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?