missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ Filtering Discs
Filtering discs made of porous PTFE.
Marque: Bohlender™ N1565-18
Informations supplémentaires : Peso : 0.09000kg
Description
- Made of porous PTFE
- Thickness: 1.0 mm
Spécification
PTFE | |
1735 mm2 | |
0.04 in. | |
47 mm |
10 μm | |
10 Pk. | |
1 mm |