CAS RN 10029-04-6
IUPAC Nome:
ethyl 2-(hydroxymethyl)prop-2-enoate
Sinonimi:
ethyl 2-(hydroxymethyl)prop-2-enoate
Peso molecolare (g/mol):
130.14
Formula molecolare:
C6H10O3
InChi Key:
SYGAXBISYRORDR-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=C)CO
1
–
3
di
3
risultati
1
Ethyl 2-(Hydroxymethyl)acrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethyl 2-(Hydroxymethyl)acrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 2-(Hydroxymethyl)acrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.