CAS RN 10031-93-3
IUPAC Nome:
ethyl 4-phenylbutanoate
Sinonimi:
ethyl 4-phenylbutanoate
Peso molecolare (g/mol):
192.26
Formula molecolare:
C12H16O2
InChi Key:
GGFNXKFGVQQNRV-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCC1=CC=CC=C1
1
–
2
di
2
risultati
1
Benzenebutanoic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzenebutanoic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.