CAS RN 100367-40-6
IUPAC Nome:
2-amino-5-bromo-4-methyl-3-nitropyridin-1-ium
Sinonimi:
2-amino-5-bromo-4-methyl-3-nitropyridin-1-ium
Peso molecolare (g/mol):
233.04
Formula molecolare:
C6H7BrN3O2
InChi Key:
KFVGEPWMVKZPND-UHFFFAOYSA-O
SMILES:
CC1=C(Br)C=[NH+]C(N)=C1[N+]([O-])=O
1
–
3
di
3
risultati
1
2-Amino-5-bromo-4-methyl-3-nitropyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-5-bromo-4-methyl-3-nitropyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Amino-5-bromo-4-methyl-3-nitropyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.