CAS RN 1005504-62-0
CAS RN 1005504-62-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
RG3039, MedChemExpress
MedChemExpress RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.