CAS RN 1006683-96-0
CAS RN 1006683-96-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
3,8-Dihydroxy-9-methoxy-6H-Dibenzo[b,d]pyran-6-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.