CAS RN 1006894-79-6
CAS RN 1006894-79-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
4-Methyl-2-(phenylmethylene)hydrazide Benzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methyl-2-(phenylmethylene)hydrazide Benzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methyl-2-(phenylmethylene)hydrazide Benzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.