CAS RN 100939-96-6
CAS RN 100939-96-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
1-(2-Methoxyphenyl)piperazine Hydrobromide, TRC
CAS: 100939-96-6 Formula molecolare: C11H16N2O . HBr Molecular Weight (g/mol): 273.17 IUPAC Name: 1-(2-methoxyphenyl)piperazine;hydrobromide SMILES: Br.COc1ccccc1N2CCNCC2