CAS RN 10108-86-8
IUPAC Nome:
trimethyl(octyl)azanium chloride
Sinonimi:
octyltrimethylammonium chloride
Peso molecolare (g/mol):
207.79
Formula molecolare:
C11H26ClN
InChi Key:
AQZSPJRLCJSOED-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCC[N+](C)(C)C
1
–
1
di
1
risultati
1
Trimethyloctylammonium Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.