CAS RN 1011244-68-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
TFAP, MedChemExpress
MedChemExpress TFAP is a selective cyclooxygenase-1 (COX-1) inhibitor, with an IC50 of 0.8 μM.