CAS RN 1020253-81-9
CAS RN 1020253-81-9
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
4'-Bromo-2'-hydroxy-5'-methylacetophenone, 97%, Thermo Scientific Chemicals
CAS: 1020253-81-9 Formula molecolare: C9H9BrO2 Molecular Weight (g/mol): 229.073 Numero MDL: MFCD08273792 InChI Key: BPQABGQSVOUODJ-UHFFFAOYSA-N Sinonimo: 1-4-bromo-2-hydroxy-5-methylphenyl ethanone,2-acetyl-4-methyl-5-bromophenol,4'-bromo-2'-hydroxy-5'-methylacetophenone,1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one,1-acetyl-4-bromo-2-hydroxy-5-methylbenzene,4'-bromo-2'-hydroxy-5'-methyl acetophenone PubChem CID: 42553093 IUPAC Name: 1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone SMILES: CC1=C(C=C(C(=C1)C(=O)C)O)Br