CAS RN 1022094-03-6
IUPAC Nome:
3-methylmorpholine hydrochloride
Sinonimi:
3-methylmorpholine hydrochloride
Peso molecolare (g/mol):
137.61
Formula molecolare:
C5H12ClNO
InChi Key:
MSOCQCWIEBVSLF-UHFFFAOYNA-N
SMILES:
Cl.CC1COCCN1
1
–
1
di
1
risultati
1
(S)-3-Methylmorpholine hydrochloride, 97%, Thermo Scientific™
CAS: 1022094-03-6 Formula molecolare: C5H11NO·ClH Molecular Weight (g/mol): 137.61 InChI Key: MSOCQCWIEBVSLF-JEDNCBNOSA-N Sinonimo: s-3-methylmorpholine hydrochloride,s-3-methylmorpholine hcl,3s-3-methylmorpholine hydrochloride,s-3-methyl-morpholine hydrochloride,s-3-methyl morpholine hydrochloride,c5h11no.clh,s-3-methylmorpholinehydrochloride,s-3-methylmorpholin hydrochloride,3s-3-methylmorpholine-hydrogen chloride 1/1 PubChem CID: 53486469 IUPAC Name: (3S)-3-methylmorpholine;hydrochloride SMILES: CC1COCCN1.Cl