CAS RN 102735-58-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
(9S,12R,13S)-Pinellic Acid, TRC
CAS: 102735-58-0 Formula molecolare: C18H34O5 Molecular Weight (g/mol): 330.47 Sinonimo: (9S,10E,12R,13S)-9,12,13-Trihydroxy-10-octadecenoic Acid,[9S-(9R*,10E,12S*,13R*)]-9,12,13-Trihydroxy-10-Octadecenoic Acid IUPAC Name: (E,9S,12R,13S)-9,12,13-trihydroxyoctadec-10-enoic acid SMILES: CCCCC[C@H](O)[C@H](O)\C=C\[C@@H](O)CCCCCCCC(=O)O