CAS RN 104776-74-1
CAS RN 104776-74-1
IUPAC Nome:
ethyl 5-(2-bromoacetyl)-1,2-oxazole-3-carboxylate
Sinonimi:
ethyl 5-(2-bromoacetyl)-1,2-oxazole-3-carboxylate
Peso molecolare (g/mol):
262.06
Formula molecolare:
C8H8BrNO4
InChi Key:
GUYZRQOTADGHII-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NOC(=C1)C(=O)CBr
1
–
1
di
1
risultati
1
ethyle5-(2-bromoacetyl)isoxazole-3-carboxylate, 97%, Thermo Scientific™
CAS: 104776-74-1 Formula molecolare: C8H8BrNO4 Molecular Weight (g/mol): 262.06 Numero MDL: MFCD00173773 InChI Key: GUYZRQOTADGHII-UHFFFAOYSA-N Sinonimo: ethyl 5-2-bromoacetyl isoxazole-3-carboxylate,ethyl 5-bromoacetyl isoxazole-3-carboxylate,ethyl 5-2-bromoacetyl-1,2-oxazole-3-carboxylate,ethyl 5-bromoacetyl-1,2-oxazole-3-carboxylate,3-isoxazolecarboxylicacid, 5-2-bromoacetyl-, ethyl ester,acmc-20a3va,5-bromoacetyl-3-ethoxycarbonyl isoxazole,ethyl 5-2-bromanylethanoyl-1,2-oxazole-3-carboxylate,5-2-bromo-1-oxoethyl-3-isoxazolecarboxylic acid ethyl ester PubChem CID: 2736373 IUPAC Name: ethyl 5-(2-bromoacetyl)-1,2-oxazole-3-carboxylate SMILES: CCOC(=O)C1=NOC(=C1)C(=O)CBr