CAS RN 109276-34-8
IUPAC Nome:
5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]-N-[5-(hydrazinecarbonyl)pentyl]pentanamide
Sinonimi:
5-[(3aS,4S,6aR)-2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[5-(hydrazinecarbonyl)pentyl]pentanamide
Peso molecolare (g/mol):
371.5
Formula molecolare:
C16H29N5O3S
InChi Key:
IJJWOSAXNHWBPR-HUBLWGQQSA-N
SMILES:
NNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
1
–
1
di
1
risultati
1
MedChemExpress Biotin LC hydrazide
MedChemExpress Biotin LC hydrazide is a long chain protein modification reagent, which can transform periodate-oxidized glycoproteins.