CAS RN 1189097-36-6
IUPAC Nome:
potassium trifluoro(3-iodophenyl)boranuide
Sinonimi:
potassium trifluoro(3-iodophenyl)boranuide
Peso molecolare (g/mol):
309.91
Formula molecolare:
C6H4BF3IK
InChi Key:
OREURDWJZUTVFM-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC(I)=CC=C1
1
–
1
di
1
risultati
1
Potassium 3-iodophenyltrifluoroborate, 96%, Thermo Scientific Chemicals
CAS: 1189097-36-6 Formula molecolare: C6H4BF3IK Molecular Weight (g/mol): 309.91 Numero MDL: MFCD09800739 InChI Key: OREURDWJZUTVFM-UHFFFAOYSA-N Sinonimo: potassium 3-iodophenyltrifluoroborate,potassium trifluoro 3-iodophenyl boranuide,amtb082 PubChem CID: 46738067 IUPAC Name: potassium;trifluoro-(3-iodophenyl)boranuide SMILES: [K+].F[B-](F)(F)C1=CC(I)=CC=C1