CAS RN 12005-88-8
CAS RN 12005-88-8
IUPAC Nome:
ferrio-λ²-arsanyl iron
Sinonimi:
ferrio-λ²-arsanyl iron
Peso molecolare (g/mol):
186.61
Formula molecolare:
AsFe2
InChi Key:
FDKAYGUKROYPRO-UHFFFAOYSA-N
SMILES:
[Fe].[Fe]=[As]
1
–
1
di
1
risultati
1
Iron arsenide, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 12005-88-8 Formula molecolare: AsFe2 Molecular Weight (g/mol): 186.612 Numero MDL: MFCD00151533 InChI Key: FDKAYGUKROYPRO-UHFFFAOYSA-N Sinonimo: 1-arsa-2,3-diferracycloprop-1-ene,iron arsenide, pieces trace metals basis PubChem CID: 71310806 SMILES: [Fe].[Fe]=[As]