CAS RN 137908-78-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Diphenhydramine N-Beta-D-Glucuronide, TRC
CAS: 137908-78-2 Formula molecolare: C23 H29 N O7 Molecular Weight (g/mol): 431.48 Sinonimo: Ethanaminium, 2-(diphenylmethoxy)-N-β-D-glucopyranuronosyl-N,N-dimethyl-, inner salt,Diphenhydramine N-glucuronide,Diphenhydramine N-β-D-Glucuronide IUPAC Name: (2S,3S,4S,5R,6R)-6-[2-benzhydryloxyethyl(dimethyl)azaniumyl]-3,4,5-trihydroxyoxane-2-carboxylate SMILES: C[N+](C)(CCOC(c1ccccc1)c2ccccc2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]