CAS RN 143782-23-4
IUPAC Nome:
4-isothiocyanato-2-(trifluoromethyl)benzonitrile
Sinonimi:
4-isothiocyanato-2-(trifluoromethyl)benzonitrile
Peso molecolare (g/mol):
228.19
Formula molecolare:
C9H3F3N2S
InChi Key:
TYXKOMAQTWRDCR-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=C(C=CC(=C1)N=C=S)C#N
1
–
2
di
2
risultati
1
4-Cyano-3-trifluoromethylphenylisothiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Cyano-3-trifluoromethylphenylisothiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.