CAS RN 15145-69-4
IUPAC Nome:
1-chloro-3-(dichloromethyl)benzene
Sinonimi:
1-chloro-3-(dichloromethyl)benzene
Peso molecolare (g/mol):
195.47
Formula molecolare:
C7H5Cl3
InChi Key:
ACNQJGLSENYFQJ-UHFFFAOYSA-N
SMILES:
ClC(Cl)C1=CC=CC(Cl)=C1
1
–
1
di
1
risultati
1
3-Chlorobenzal chloride, 95%, Thermo Scientific™
CAS: 15145-69-4 Formula molecolare: C7H5Cl3 Molecular Weight (g/mol): 195.467 Numero MDL: MFCD00082691 InChI Key: ACNQJGLSENYFQJ-UHFFFAOYSA-N PubChem CID: 84803 IUPAC Name: 1-chloro-3-(dichloromethyl)benzene SMILES: C1=CC(=CC(=C1)Cl)C(Cl)Cl