CAS RN 177787-26-7
IUPAC Nome:
3,4,5-trifluorobenzoyl chloride
Sinonimi:
3,4,5-trifluorobenzoyl chloride
Peso molecolare (g/mol):
194.54
Formula molecolare:
C7H2ClF3O
InChi Key:
YRUNCQNKZIQTEO-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC(F)=C1F)C(Cl)=O
1
–
1
di
1
risultati
1
3,4,5-Trifluorobenzoyl chloride, 97%, Thermo Scientific™
CAS: 177787-26-7 Formula molecolare: C7H2ClF3O Molecular Weight (g/mol): 194.537 Numero MDL: MFCD00083526 InChI Key: YRUNCQNKZIQTEO-UHFFFAOYSA-N Sinonimo: 3,4,5-trifluorobenzoylchloride,benzoyl chloride,3,4,5-trifluoro,acmc-20ange,pubchem20778,3,4,5-trifluorobenzoic acid chloride,3,4,5-tris fluoranyl benzoyl chloride,3,4,5-trifluorobenzoyl chloride,benzoyl chloride, 3,4,5-trifluoro-9ci PubChem CID: 2777023 IUPAC Name: 3,4,5-trifluorobenzoyl chloride SMILES: C1=C(C=C(C(=C1F)F)F)C(=O)Cl