CAS RN 188781-46-6
IUPAC Nome:
2,4-bis(trifluoromethyl)pyrimidine-5-carboxylic acid
Sinonimi:
2,4-bis(trifluoromethyl)pyrimidine-5-carboxylic acid
Peso molecolare (g/mol):
260.1
Formula molecolare:
C7H2F6N2O2
InChi Key:
RUHKDOFTFCXTBX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN=C(N=C1C(F)(F)F)C(F)(F)F