CAS RN 19829-79-9
IUPAC Nome:
8-hydroxyquinoline-7-carboxylic acid
Sinonimi:
8-hydroxyquinoline-7-carboxylic acid
Peso molecolare (g/mol):
189.17
Formula molecolare:
C10H7NO3
InChi Key:
JYIAZVFJRYLCBH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C2C=CC=NC2=C1O
1
–
1
di
1
risultati
1
8-Hydroxyquinoline-7-carboxylic Acid, TRC
CAS: 19829-79-9 Formula molecolare: C10H7NO3 Molecular Weight (g/mol): 189.16 Sinonimo: 7-Carboxy-8-hydroxyquinoline,NSC 27445,NSC 35082 IUPAC Name: 8-hydroxyquinoline-7-carboxylic acid SMILES: OC(=O)c1ccc2cccnc2c1O