CAS RN 200958-40-3
IUPAC Nome:
2-bromo-4-nitro-1-(trifluoromethoxy)benzene
Sinonimi:
2-bromo-4-nitro-1-(trifluoromethoxy)benzene
Peso molecolare (g/mol):
286
Formula molecolare:
C7H3BrF3NO3
InChi Key:
LVTAFGFYMLODQP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(Br)=C(OC(F)(F)F)C=C1
1
–
1
di
1
risultati
1
2-Bromo-4-nitro-1-(trifluoromethoxy)benzene, 98%
CAS: 200958-40-3 Formula molecolare: C7H3BrF3NO3 Molecular Weight (g/mol): 286.00 Numero MDL: MFCD04973758 InChI Key: LVTAFGFYMLODQP-UHFFFAOYSA-N PubChem CID: 24721641 IUPAC Name: 2-bromo-4-nitro-1-(trifluoromethoxy)benzene SMILES: [O-][N+](=O)C1=CC(Br)=C(OC(F)(F)F)C=C1