CAS RN 20458-99-5
IUPAC Nome:
1,3-diethyl-2-isocyanatobenzene
Sinonimi:
1,3-diethyl-2-isocyanatobenzene
Peso molecolare (g/mol):
175.23
Formula molecolare:
C11H13NO
InChi Key:
AUSRFKVPUCQCCN-UHFFFAOYSA-N
SMILES:
CCC1=CC=CC(CC)=C1N=C=O
1
–
1
di
1
risultati
1
2,6-Diethylphenyl isocyanate, 98+%
CAS: 20458-99-5 Formula molecolare: C11H13NO Molecular Weight (g/mol): 175.23 Numero MDL: MFCD00019912 InChI Key: AUSRFKVPUCQCCN-UHFFFAOYSA-N Sinonimo: 2,6-diethylphenyl isocyanate,2,6-diethylphenylisocyanate,1,3-diethyl-2-isocyanato-benzene,benzene, 1,3-diethyl-2-isocyanato,acmc-1co7s,2,6-diethylbenzenisocyanate,benzene,1,3-diethyl-2-isocyanato PubChem CID: 5153183 IUPAC Name: 1,3-diethyl-2-isocyanatobenzene SMILES: CCC1=CC=CC(CC)=C1N=C=O