CAS RN 21450-90-8
IUPAC Nome:
magnesium(2+) 2-methylnaphthalen-1-ide bromide
Sinonimi:
magnesium(2+) 2-methylnaphthalen-1-ide bromide
Peso molecolare (g/mol):
245.4
Formula molecolare:
C11H9BrMg
InChi Key:
LYRLLQRVTWRTAJ-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CC1=CC=C2C=CC=CC2=[C-]1