CAS RN 2250019-92-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
R&D Systems™ AMG 18 hydrochloride
A Tocris product, part of R&D Systems. IRE1 Inhibitors
2