CAS RN 24623-20-9
IUPAC Nome:
6-methyl-2,3-dihydro-1H-inden-1-one
Sinonimi:
6-methyl-2,3-dihydroinden-1-one
Peso molecolare (g/mol):
146.19
Formula molecolare:
C10H10O
InChi Key:
DBOXRDYLMJMQBB-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(CCC2=O)C=C1
1
–
1
di
1
risultati
1
6-Methyl-1-indanone, 98%, Thermo Scientific™
CAS: 24623-20-9 Formula molecolare: C10H10O Molecular Weight (g/mol): 146.19 Numero MDL: MFCD00275729 InChI Key: DBOXRDYLMJMQBB-UHFFFAOYSA-N Sinonimo: 6-methyl-1-indanone,6-methyl-2,3-dihydro-1h-inden-1-one,6-methylindan-1-one,6-methyl-indan-1-one,1h-inden-1-one, 2,3-dihydro-6-methyl,2,3-dihydro-6-methylinden-1-one,pubchem8851,acmc-209gdm,6-methyl-indane-1-one PubChem CID: 2795093 IUPAC Name: 6-methyl-2,3-dihydroinden-1-one SMILES: CC1=CC2=C(CCC2=O)C=C1