CAS RN 30614-77-8
IUPAC Nome:
3,4-diethyl furan-3,4-dicarboxylate
Sinonimi:
3,4-diethyl furan-3,4-dicarboxylate
Peso molecolare (g/mol):
212.2
Formula molecolare:
C10H12O5
InChi Key:
ZODFWNHYQARJLC-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=COC=C1C(=O)OCC