CAS RN 319-86-8
CAS RN 319-86-8
IUPAC Nome:
1,2,3,4,5,6-hexachlorocyclohexane
Sinonimi:
lindane hexachlorocyclohexane 林旦 hexachlorocyclohexanes hexachlorzyklohexan hexachlorcyclohexan benzene hexachloride
Peso molecolare (g/mol):
290.81
Formula molecolare:
C6H6Cl6
InChi Key:
JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES:
ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl