CAS RN 34730-20-6
IUPAC Nome:
1-(4-chlorothiophen-2-yl)ethan-1-one
Sinonimi:
1-(4-chlorothiophen-2-yl)ethanone
Peso molecolare (g/mol):
160.62
Formula molecolare:
C6H5ClOS
InChi Key:
FKESGQASARHBDC-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Cl)=CS1
1
–
1
di
1
risultati
1
2-Acetyl-4-chlorothiophene, 98+%
CAS: 34730-20-6 Formula molecolare: C6H5ClOS Molecular Weight (g/mol): 160.62 Numero MDL: MFCD00082791 InChI Key: FKESGQASARHBDC-UHFFFAOYSA-N PubChem CID: 11105655 IUPAC Name: 1-(4-chlorothiophen-2-yl)ethanone SMILES: CC(=O)C1=CC(Cl)=CS1