CAS RN 37928-17-9
IUPAC Nome:
5-methyl-3,4-diphenyl-1,2-oxazole
Sinonimi:
5-methyl-3,4-diphenyl-1,2-oxazole
Peso molecolare (g/mol):
235.29
Formula molecolare:
C16H13NO
InChi Key:
ZXIRUKJWLADSJS-UHFFFAOYSA-N
SMILES:
CC1=C(C(=NO1)C1=CC=CC=C1)C1=CC=CC=C1