CAS RN 38083-17-9
IUPAC Nome:
1-(4-chlorophenoxy)-1-(1H-imidazol-1-yl)-3,3-dimethylbutan-2-one
Sinonimi:
climbazole 氯咪巴唑
Peso molecolare (g/mol):
292.76
Formula molecolare:
C15H17ClN2O2
InChi Key:
OWEGWHBOCFMBLP-UHFFFAOYNA-N
SMILES:
CC(C)(C)C(=O)C(OC1=CC=C(Cl)C=C1)N1C=CN=C1
1
–
2
di
2
risultati
1
Climbazole, TRC
CAS: 38083-17-9 Formula molecolare: C15 H17 Cl N2 O2 Molecular Weight (g/mol): 292.76 Sinonimo: 1-(4-Chlorophenoxy)-1-(1H-imidazol-1-yl)-3,3-dimethyl-2-butanone,1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one,1-[(4-Chlorophenoxy)(tert-butylcarbonyl)methyl]imidazole,BAY-e 6975,Baypival,Climbazole,Crinipan AD,Diazolone IUPAC Name: 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one SMILES: CC(C)(C)C(=O)C(Oc1ccc(Cl)cc1)n2ccnc2
2
Climbazole, MedChemExpress
MedChemExpress Climbazole (BAY-e 6975) is a potent antifungal agent. Climbazole also is a potent inducer of rat hepatic cytochrome P450.