CAS RN 406-86-0
IUPAC Nome:
4,4,4-trifluorobut-2-enenitrile
Sinonimi:
4,4,4-trifluorobut-2-enenitrile
Peso molecolare (g/mol):
121.06
Formula molecolare:
C4H2F3N
InChi Key:
LHWSEFCIRYVTLZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C=CC#N