CAS RN 481-74-3
IUPAC Nome:
1,8-dihydroxy-3-methyl-9,10-dihydroanthracene-9,10-dione
Sinonimi:
turkey rhubarb C.I. natural yellow 23 chrysophanol chrysophanic acid 3-methylchrysazin chrysophansaeure
Peso molecolare (g/mol):
254.24
Formula molecolare:
C15H10O4
InChi Key:
LQGUBLBATBMXHT-UHFFFAOYSA-N
SMILES:
CC1=CC(O)=C2C(=O)C3=C(O)C=CC=C3C(=O)C2=C1
1
–
3
di
3
risultati
1
Chrysophanic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Chrysophanic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Chrysophanol, MedChemExpress
MedChemExpress Chrysophanol (Chrysophanic acid) is a natural anthraquinone, which inhibits EGF-induced phosphorylation of EGFR and suppresses activation of AKT and mTOR/p70S6K.