CAS RN 5002-47-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Fluphenazine Decanoate (~90%), TRC
CAS: 5002-47-1 Formula molecolare: C32 H44 F3 N3 O2 S Molecular Weight (g/mol): 591.77 Sinonimo: Fluphenazine decanoate,2-[4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]piperazin-1-yl]ethyl decanoate IUPAC Name: 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethyl decanoate SMILES: CCCCCCCCCC(=O)OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1