CAS RN 52206-05-0
IUPAC Nome:
1-acetyl-2,3-dihydro-1H-indole-5-sulfonyl chloride
Sinonimi:
1-acetyl-2,3-dihydroindole-5-sulfonyl chloride
Peso molecolare (g/mol):
259.7
Formula molecolare:
C10H10ClNO3S
InChi Key:
QNFXLCHANYHGIF-UHFFFAOYSA-N
SMILES:
CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O
1
–
1
di
1
risultati
1
1-Acetylindoline-5-sulfonyl chloride, 97%
CAS: 52206-05-0 Formula molecolare: C10H10ClNO3S Molecular Weight (g/mol): 259.70 Numero MDL: MFCD07368558 InChI Key: QNFXLCHANYHGIF-UHFFFAOYSA-N Sinonimo: 1-acetyl-2,3-dihydro-1h-indole-5-sulfonyl chloride,1-acetylindoline-5-sulfonyl chloride,1-acetyl-5-indolinesulfonoyl chloride,1h-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro,1-acetyl-5-indolinesulfonyl chloride,n-acetylindoline-5-sulphonyl chloride,1-acetyl-5-chlorosulfonyl indoline,1-acetylindoline-5-sulfonylchloride,pubchem5449,acetylindolinesulfonoylchloride PubChem CID: 14024596 IUPAC Name: 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride SMILES: CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O