CAS RN 609-26-7
IUPAC Nome:
3-ethyl-2-methylpentane
Sinonimi:
2-methyl-3-ethylpentane
Peso molecolare (g/mol):
114.23
Formula molecolare:
C8H18
InChi Key:
DUPUVYJQZSLSJB-UHFFFAOYSA-N
SMILES:
CCC(CC)C(C)C
1
–
3
di
3
risultati
1
3-Ethyl-2-methylpentane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Ethyl-2-methylpentane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Ethyl-2-methylpentane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.