CAS RN 65944-23-2
IUPAC Nome:
bis(acridin-9-amine) hydrate
Sinonimi:
bis(9-aminoacridine) hydrate
Peso molecolare (g/mol):
406.49
Formula molecolare:
C26H22N4O
InChi Key:
SPWFQHCZHQCDAG-UHFFFAOYSA-N
SMILES:
O.NC1=C2C=CC=CC2=NC2=CC=CC=C12.NC1=C2C=CC=CC2=NC2=CC=CC=C12